Assay Details
Target Gene Details
Entrez Gene ID: | 116159 |
Gene Name: | cysteine and tyrosine rich 1 |
Gene Aliases: |
C21orf95 |
Location: |
Chr.21:26466209-26573404 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYYR1 | NM_001320768.1 | NP_001307697.1 | ||
| NM_052954.3 | NP_443186.2 | |||
| NR_135472.1 | ||||
| XM_011529450.2 | XP_011527752.1 | |||
| AF401639.1 | AAK84158.1 | |||
| AK054581.1 | BAB70764.1 | |||
| AK223576.1 | BAD97296.1 | |||
| AK304124.1 | ||||
| AK313231.1 | ||||
| AL833200.1 | CAI46188.1 | |||
| AY061853.1 | AAL35737.1 | |||
| BC036761.1 | AAH36761.1 | |||
| JF693912.1 | ||||
| JF693913.1 | ||||
| JF693914.1 | ||||
| KC843341.1 | ||||
| KF154022.1 |
Target Gene Details
Entrez Gene ID: | 100996571 |
Gene Name: | cysteine and tyrosine rich 1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.21:26393635-26569252 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYYR1-AS1 | NR_135515.1 | |||
| NR_135516.1 | ||||
| AK125338.1 | BAC86136.1 | |||
| BX114294.1 | ||||
| JF693915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587336 | Chr.21:26477027 - 26493659 on Build GRCh38 | Loss |
|
| nsv3481 | Chr.21:26460916 - 26495488 on Build GRCh38 | Insertion |
|
| nsv587335 | Chr.21:26342944 - 26583066 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map