Assay Details
Target Gene Details
Entrez Gene ID: | 2114 |
Gene Name: | ETS proto-oncogene 2, transcription factor |
Gene Aliases: |
ETS2IT1 |
Location: |
Chr.21:38805307-38824955 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETS2 | NM_001256295.1 | NP_001243224.1 | ||
| NM_005239.5 | NP_005230.1 | |||
| AK096841.1 | ||||
| AK315563.1 | ||||
| BC017040.1 | AAH17040.1 | |||
| BC042954.2 | AAH42954.1 | |||
| J04102.1 | AAA52412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058591 | Chr.21:38742206 - 38818670 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map