Assay Details
Target Gene Details
Entrez Gene ID: | 10600 |
Gene Name: | ubiquitin specific peptidase 16 |
Gene Aliases: |
UBP-M, UBPM |
Location: |
Chr.21:29024617-29054488 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP16 | NM_001001992.1 | NP_001001992.1 | ||
| NM_001032410.1 | NP_001027582.1 | |||
| NM_006447.2 | NP_006438.1 | |||
| XM_017028257.1 | XP_016883746.1 | |||
| XM_017028258.1 | XP_016883747.1 | |||
| XM_017028259.1 | XP_016883748.1 | |||
| XM_017028261.1 | XP_016883750.1 | |||
| AF126736.1 | AAD20949.1 | |||
| AK025104.1 | ||||
| AK025989.1 | ||||
| AK222681.1 | BAD96401.1 | |||
| AK222884.1 | BAD96604.1 | |||
| AK294284.1 | ||||
| AK300771.1 | ||||
| AY333928.1 | AAR13293.1 | |||
| BC030777.2 | AAH30777.1 | |||
| CB960929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893402 | Chr.21:28969569 - 29059824 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map