Assay Details
Target Gene Details
Entrez Gene ID: | 59271 |
Gene Name: | eva-1 homolog C (C. elegans) |
Gene Aliases: |
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21 |
Location: |
Chr.21:32411692-32515400 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVA1C | NM_001320745.1 | 1 | 303 | NP_001307674.1 |
| XM_017028420.1 | 1 | 269 | XP_016883909.1 | |
| XM_017028421.1 | 1 | 303 | XP_016883910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834085 | Chr.21:32322748 - 32519107 on Build GRCh38 | Gain |
|
| nsv3494 | Chr.21:32401757 - 32436315 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map