Assay Details
Target Gene Details
Entrez Gene ID: | 1525 |
Gene Name: | coxsackie virus and adenovirus receptor |
Gene Aliases: |
CAR, CAR4/6, HCAR |
Location: |
Chr.21:17512906-17636213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXADR | NM_001207063.1 | NP_001193992.1 | ||
| NM_001207064.1 | NP_001193993.1 | |||
| NM_001207065.1 | NP_001193994.1 | |||
| NM_001207066.1 | NP_001193995.1 | |||
| NM_001338.4 | NP_001329.1 | |||
| XM_011529476.2 | XP_011527778.1 | |||
| XM_011529477.2 | XP_011527779.1 | |||
| XM_011529478.2 | XP_011527780.1 | |||
| XM_011529479.1 | XP_011527781.1 | |||
| AF124598.1 | AAD31772.1 | |||
| AK313526.1 | ||||
| AY072910.1 | AAL68878.1 | |||
| AY072911.1 | AAL68879.1 | |||
| AY072912.1 | AAL68880.1 | |||
| BC003684.1 | AAH03684.1 | |||
| BC010536.1 | AAH10536.1 | |||
| BM547121.1 | ||||
| BT019876.1 | AAV38679.1 | |||
| CV570985.1 | ||||
| GU474812.1 | ||||
| U90716.1 | AAC51234.1 | |||
| Y07593.1 | CAA68868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575381 | Chr.21:17436662 - 17709158 on Build GRCh38 | Gain |
|
| esv3893382 | Chr.21:17364904 - 17698240 on Build GRCh38 | Gain |
|
| esv2723192 | Chr.21:16398054 - 17697109 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map