Assay Details
Target Gene Details
Entrez Gene ID: | 9514 |
Gene Name: | galactose-3-O-sulfotransferase 1 |
Gene Aliases: |
CST |
Location: |
Chr.22:30554635-30574588 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAL3ST1 | NM_001318103.1 | NP_001305032.1 | ||
| NM_001318104.1 | NP_001305033.1 | |||
| NM_001318105.1 | NP_001305034.1 | |||
| NM_001318106.1 | NP_001305035.1 | |||
| NM_001318107.1 | NP_001305036.1 | |||
| NM_001318108.1 | NP_001305037.1 | |||
| NM_001318109.1 | NP_001305038.1 | |||
| XM_011530518.2 | XP_011528820.1 | |||
| XM_011530522.2 | XP_011528824.1 | |||
| XM_011530524.1 | XP_011528826.1 | |||
| XM_017029097.1 | 1 | 95 | XP_016884586.1 | |
| XM_017029098.1 | 1 | 94 | XP_016884587.1 | |
| AK313097.1 | ||||
| BM923857.1 | ||||
| BP220526.1 | ||||
| DA271385.1 | ||||
| DA372454.1 | ||||
| DA376354.1 | ||||
| DA382951.1 | ||||
| DA384904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588887 | Chr.22:30557534 - 30599369 on Build GRCh38 | Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map