Assay Details
Target Gene Details
Entrez Gene ID: | 26286 |
Gene Name: | ADP ribosylation factor GTPase activating protein 3 |
Gene Aliases: |
ARFGAP1 |
Location: |
Chr.22:42796526-42857402 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFGAP3 | NM_001142293.1 | NP_001135765.1 | ||
| NM_014570.4 | NP_055385.3 | |||
| XM_005261525.3 | XP_005261582.1 | |||
| AF111847.1 | AAF40310.1 | |||
| AK002083.1 | BAA92076.1 | |||
| AK022768.1 | BAB14236.1 | |||
| AK127525.1 | ||||
| AK297635.1 | ||||
| AK300531.1 | ||||
| AK307967.1 | ||||
| AK315720.1 | ||||
| AL137598.1 | CAB70834.1 | |||
| AL159143.1 | CAB76901.1 | |||
| BC005122.2 | AAH05122.1 | |||
| CR456382.1 | CAG30268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589084 | Chr.22:42776261 - 42807131 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map