Assay Details
Target Gene Details
Entrez Gene ID: | 57168 |
Gene Name: | aspartate beta-hydroxylase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.22:26429314-26445012 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPHD2 | NM_020437.4 | NP_065170.2 | ||
| AK097157.1 | ||||
| AK315112.1 | ||||
| AL161993.1 | CAB82325.1 | |||
| BC036753.1 | AAH36753.1 | |||
| CR456391.1 | CAG30277.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114539770,rs76631461] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map