Assay Details
Target Gene Details
Entrez Gene ID: | 11252 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 2 |
Gene Aliases: |
SDPII |
Location: |
Chr.22:42869766-43015690 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508738 | Chr.22:42839659 - 42917996 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map