Assay Details
Target Gene Details
Entrez Gene ID: | 25771 |
Gene Name: | TBC1 domain family member 22A |
Gene Aliases: |
C22orf4, HSC79E021 |
Location: |
Chr.22:46762617-47224975 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3654 | Chr.22:46755700 - 46775645 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map