Assay Details
Target Gene Details
Entrez Gene ID: | 2547 |
Gene Name: | X-ray repair cross complementing 6 |
Gene Aliases: |
CTC75, CTCBF, G22P1, KU70, ML8, TLAA |
Location: |
Chr.22:41621163-41664048 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XRCC6 | NM_001288976.1 | NP_001275905.1 | ||
| NM_001288977.1 | NP_001275906.1 | |||
| NM_001288978.1 | NP_001275907.1 | |||
| NM_001469.4 | NP_001460.1 | |||
| AK055786.1 | ||||
| AK293439.1 | ||||
| AK304580.1 | ||||
| AK315618.1 | ||||
| BC008343.1 | AAH08343.1 | |||
| BC010034.2 | AAH10034.1 | |||
| BC012154.2 | AAH12154.1 | |||
| BC018259.2 | AAH18259.1 | |||
| BC072449.1 | AAH72449.1 | |||
| BQ217843.1 | ||||
| BU902323.1 | ||||
| CR456492.1 | CAG30378.1 | |||
| CR542219.1 | CAG47015.1 | |||
| J04607.1 | AAA61177.1 | |||
| J04611.1 | AAA51733.1 | |||
| M32865.1 | AAA36155.1 | |||
| S38729.1 | AAB22381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv966111 | Chr.22:41634907 - 41640750 on Build GRCh38 | Duplication |
|
| nsv834209 | Chr.22:41563516 - 41746056 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map