Assay Details
Target Gene Details
Entrez Gene ID: | 8563 |
Gene Name: | THO complex 5 |
Gene Aliases: |
C22orf19, Fmip, PK1.3, fSAP79 |
Location: |
Chr.22:29508167-29554254 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THOC5 | NM_001002877.1 | NP_001002877.1 | ||
| NM_001002878.1 | NP_001002878.1 | |||
| NM_001002879.1 | NP_001002879.1 | |||
| NM_003678.4 | NP_003669.4 | |||
| XM_005261797.1 | XP_005261854.1 | |||
| XM_005261798.1 | XP_005261855.1 | |||
| XM_005261799.1 | XP_005261856.1 | |||
| XM_005261801.3 | XP_005261858.1 | |||
| XM_006724347.1 | XP_006724410.1 | |||
| XM_017029027.1 | XP_016884516.1 | |||
| AB023200.1 | ||||
| AK025385.1 | ||||
| AK098709.1 | ||||
| AK122673.1 | ||||
| AK225752.1 | ||||
| BC003615.2 | AAH03615.1 | |||
| BQ184247.1 | ||||
| BX400943.2 | ||||
| CR456542.1 | CAG30428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834172 | Chr.22:29503047 - 29661959 on Build GRCh38 | Loss |
|
| esv3647538 | Chr.22:29506399 - 29525757 on Build GRCh38 | Gain |
|
| esv3647536 | Chr.22:29477608 - 29543952 on Build GRCh38 | Gain |
|
| esv3893473 | Chr.22:29474962 - 29528836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map