Assay Details
Target Gene Details
Entrez Gene ID: | 113828 |
Gene Name: | family with sequence similarity 83 member F |
Gene Aliases: |
- |
Location: |
Chr.22:39994949-40030039 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM83F | NM_138435.2 | NP_612444.2 | ||
| BC011204.1 | AAH11204.1 | |||
| BC031099.1 | AAH31099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955160 | Chr.22:40016997 - 40040396 on Build GRCh38 | Deletion |
|
| nsv834201 | Chr.22:39923541 - 40131810 on Build GRCh38 | Gain |
|

Genomic Map