Assay Details
Target Gene Details
Entrez Gene ID: | 1454 |
Gene Name: | casein kinase 1 epsilon |
Gene Aliases: |
CKIepsilon, HCKIE |
Location: |
Chr.22:38290691-38318084 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1E | NM_001894.4 | NP_001885.1 | ||
| NM_152221.2 | NP_689407.1 | |||
| AB024597.1 | BAA92345.1 | |||
| AB091043.1 | BAC10902.1 | |||
| AK223498.1 | BAD97218.1 | |||
| BC006490.2 | AAH06490.1 | |||
| BT019831.1 | AAV38634.1 | |||
| CR456429.1 | CAG30315.1 | |||
| L37043.1 | AAC41761.1 |
Target Gene Details
Entrez Gene ID: | 102800317 |
Gene Name: | LOC400927-CSNK1E readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:38290691-38398522 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400927-CSNK1E | NM_001289912.1 | NP_001276841.1 | ||
| AK092269.1 |
More Information
Set Membership: |
|

Genomic Map