Assay Details
Target Gene Details
Entrez Gene ID: | 440822 |
Gene Name: | piwi like RNA-mediated gene silencing 3 |
Gene Aliases: |
HIWI3 |
Location: |
Chr.22:24719034-24774720 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIWIL3 | NM_001008496.3 | NP_001008496.2 | ||
| NM_001255975.1 | NP_001242904.1 | |||
| NR_045648.1 | ||||
| NR_045649.1 | ||||
| AB079368.1 | BAC81343.1 | |||
| AK302409.1 | ||||
| AK315806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893464 | Chr.22:24683958 - 24728744 on Build GRCh38 | Gain |
|
| esv2611432 | Chr.22:24721171 - 24722718 on Build GRCh38 | Deletion |
|
| nsv3579 | Chr.22:24702064 - 24735980 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map