Assay Details
Target Gene Details
Entrez Gene ID: | 2130 |
Gene Name: | EWS RNA binding protein 1 |
Gene Aliases: |
EWS, EWS-FLI1, bK984G1.4 |
Location: |
Chr.22:29268009-29300525 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2669638 | Chr.22:29283419 - 29285901 on Build GRCh38 | Deletion |
|
| esv3647529 | Chr.22:29283431 - 29285907 on Build GRCh38 | Loss |
|
| esv21927 | Chr.22:29284294 - 29285816 on Build GRCh38 | Loss |
|
| nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss |
|
| nsv829154 | Chr.22:29221496 - 29328012 on Build GRCh38 | Gain |
|
| nsv829155 | Chr.22:29268407 - 29328012 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74438268] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map