Assay Details
Target Gene Details
Entrez Gene ID: | 646851 |
Gene Name: | family with sequence similarity 227 member A |
Gene Aliases: |
- |
Location: |
Chr.22:38578120-38656629 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM227A | NM_001013647.1 | 17 | 9978 | NP_001013669.1 |
| NM_001291030.1 | 17 | 10018 | NP_001277959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3624 | Chr.22:38552181 - 38602777 on Build GRCh38 | Deletion |
|

Genomic Map