Assay Details
Target Gene Details
Entrez Gene ID: | 25809 |
Gene Name: | tubulin tyrosine ligase like 1 |
Gene Aliases: |
C22orf7, HS323M22B |
Location: |
Chr.22:43039516-43089428 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL1 | NM_012263.4 | NP_036395.1 | ||
| NR_027779.1 | ||||
| XM_006724225.3 | XP_006724288.1 | |||
| XM_011530108.2 | XP_011528410.1 | |||
| XM_011530110.2 | XP_011528412.1 | |||
| XM_017028751.1 | XP_016884240.1 | |||
| XM_017028752.1 | XP_016884241.1 | |||
| XM_017028753.1 | XP_016884242.1 | |||
| AF104927.1 | AAG29879.1 | |||
| AF173935.1 | AAF91088.1 | |||
| AK095996.1 | ||||
| AK315658.1 | ||||
| AL096883.1 | CAB51423.1 | |||
| AL096886.1 | CAB51469.1 | |||
| AL136687.1 | CAB66622.1 | |||
| AL589867.1 | CAC34478.1 | |||
| BC014968.1 | AAH14968.1 | |||
| BG911167.1 | ||||
| CR456599.1 | CAG30485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762124 | Chr.22:43031130 - 43188670 on Build GRCh38 | Gain |
|
| nsv1061240 | Chr.22:43019575 - 43120127 on Build GRCh38 | Gain |
|
| nsv834214 | Chr.22:43037931 - 43217928 on Build GRCh38 | Gain |
|
| nsv3645 | Chr.22:43030260 - 43062792 on Build GRCh38 | Insertion |
|
| nsv834213 | Chr.22:42935490 - 43094181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map