Assay Details
Target Gene Details
Entrez Gene ID: | 6523 |
Gene Name: | solute carrier family 5 member 1 |
Gene Aliases: |
D22S675, NAGT, SGLT1 |
Location: |
Chr.22:32043032-32113029 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A1 | NM_000343.3 | 1 | 65 | NP_000334.1 |
| XM_011530331.1 | XP_011528633.1 | |||
| AK312948.1 | ||||
| DA429018.1 | ||||
| DQ656053.2 | 1 | 65 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4560n100 | Chr.22:32004579 - 32057060 on Build GRCh38 | Loss |
|
| esv3647597 | Chr.22:31969894 - 32151343 on Build GRCh38 | Gain |
|
| nsv834177 | Chr.22:32020692 - 32198676 on Build GRCh38 | Gain |
|
| dgv4559n100 | Chr.22:31960163 - 32072845 on Build GRCh38 | Gain |
|
| esv3647598 | Chr.22:32004592 - 32053519 on Build GRCh38 | Gain |
|
| esv3575475 | Chr.22:31962552 - 32068189 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map