Assay Details
Target Gene Details
Entrez Gene ID: | 1454 |
Gene Name: | casein kinase 1 epsilon |
Gene Aliases: |
CKIepsilon, HCKIE |
Location: |
Chr.22:38290691-38318084 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1E | NM_001894.4 | NP_001885.1 | ||
| NM_152221.2 | NP_689407.1 | |||
| AB024597.1 | BAA92345.1 | |||
| AB091043.1 | BAC10902.1 | |||
| AK223498.1 | BAD97218.1 | |||
| BC006490.2 | AAH06490.1 | |||
| BQ434269.1 | ||||
| BT019831.1 | AAV38634.1 | |||
| CF594072.1 | ||||
| CR456429.1 | CAG30315.1 | |||
| L37043.1 | AAC41761.1 |
Target Gene Details
Entrez Gene ID: | 102800317 |
Gene Name: | LOC400927-CSNK1E readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:38290691-38398522 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400927-CSNK1E | NM_001289912.1 | NP_001276841.1 | ||
| AK092269.1 | ||||
| AK310303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834195 | Chr.22:38300366 - 38490431 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map