Assay Details
Target Gene Details
Entrez Gene ID: | 3162 |
Gene Name: | heme oxygenase 1 |
Gene Aliases: |
HMOX1D, HO-1, HSP32, bK286B10 |
Location: |
Chr.22:35381067-35394214 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMOX1 | NM_002133.2 | NP_002124.1 | ||
| AK313120.1 | ||||
| BC001491.2 | AAH01491.1 | |||
| BT019785.1 | AAV38588.1 | |||
| CR456505.1 | CAG30391.1 | |||
| CR541945.1 | CAG46743.1 | |||
| CR541968.1 | CAG46766.1 | |||
| JF323038.1 | ||||
| X06985.1 | CAA30045.1 |
More Information
Set Membership: |
|

Genomic Map