Assay Details
Target Gene Details
Entrez Gene ID: | 4771 |
Gene Name: | neurofibromin 2 |
Gene Aliases: |
ACN, BANF, SCH |
Location: |
Chr.22:29603556-29698600 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834172 | Chr.22:29503047 - 29661959 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map