Assay Details
Target Gene Details
Entrez Gene ID: | 652968 |
Gene Name: | GATS protein like 3 |
Gene Aliases: |
CASTOR1 |
Location: |
Chr.22:30285118-30289627 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GATSL3 | NM_001037666.2 | NP_001032755.1 | ||
| AK055587.1 | BAB70963.1 | |||
| AK309436.1 | ||||
| AK310246.1 | ||||
| BC021927.1 | AAH21927.1 | |||
| CR456449.1 | CAG30335.1 | |||
| DA343534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588885 | Chr.22:30265195 - 30295642 on Build GRCh38 | Loss |
|
| nsv1062388 | Chr.22:30253190 - 30315454 on Build GRCh38 | Gain |
|
| nsv834174 | Chr.22:30223771 - 30350926 on Build GRCh38 | Gain+Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map