Assay Details
Target Gene Details
Entrez Gene ID: | 646851 |
Gene Name: | family with sequence similarity 227 member A |
Gene Aliases: |
- |
Location: |
Chr.22:38578120-38656629 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM227A | NM_001013647.1 | 17 | 5801 | NP_001013669.1 |
| NM_001291030.1 | 17 | 5841 | NP_001277959.1 | |
| XM_006724291.2 | 18 | 2740 | XP_006724354.2 | |
| XM_006724292.2 | 18 | 2728 | XP_006724355.2 | |
| XM_006724294.2 | 18 | 2780 | XP_006724357.2 | |
| XM_006724295.2 | 18 | 2768 | XP_006724358.2 | |
| XM_011530308.2 | 18 | 2445 | XP_011528610.1 | |
| XM_011530309.2 | XP_011528611.1 | |||
| XM_011530310.1 | 17 | 2635 | XP_011528612.1 | |
| XM_011530311.1 | 17 | 2617 | XP_011528613.1 | |
| XM_011530312.1 | 16 | 2017 | XP_011528614.1 | |
| XM_011530313.2 | 16 | 1969 | XP_011528615.1 | |
| XM_011530314.1 | 17 | 2670 | XP_011528616.1 | |
| AK095080.1 | 1 | 514 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3624 | Chr.22:38552181 - 38602777 on Build GRCh38 | Deletion |
|

Genomic Map