Assay Details
Target Gene Details
Entrez Gene ID: | 4248 |
Gene Name: | mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase |
Gene Aliases: |
GNT-III, GNT3 |
Location: |
Chr.22:39447166-39492194 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT3 | XM_005261608.4 | 1 | 38 | XP_005261665.1 |
| XM_011530191.2 | 1 | 38 | XP_011528493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471200 | Chr.22:39416404 - 39478309 on Build GRCh38 | Loss |
|
| nsv834199 | Chr.22:39324389 - 39531343 on Build GRCh38 | Loss |
|
| nsv3628 | Chr.22:39435098 - 39468570 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map