Assay Details
Target Gene Details
Entrez Gene ID: | 26088 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.22:37607235-37633564 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGA1 | XM_011530122.1 | XP_011528424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| nsv471197 | Chr.22:37600773 - 37762734 on Build GRCh38 | Loss |
|
| nsv834191 | Chr.22:37533773 - 37717344 on Build GRCh38 | Loss |
|
| nsv1057617 | Chr.22:37462633 - 37713698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map