Assay Details
Target Gene Details
Entrez Gene ID: | 400927 |
Gene Name: | TPTE and PTEN homologous inositol lipid phosphatase pseudogene |
Gene Aliases: |
- |
Location: |
Chr.22:38344665-38398926 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400927 | NR_002821.2 | |||
| AK307225.1 | ||||
| CN357790.1 |
Target Gene Details
Entrez Gene ID: | 102800317 |
Gene Name: | LOC400927-CSNK1E readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:38290691-38398522 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400927-CSNK1E | NM_001289912.1 | NP_001276841.1 | ||
| AK092269.1 | ||||
| AK310303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834195 | Chr.22:38300366 - 38490431 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map