Assay Details
Target Gene Details
Entrez Gene ID: | 25771 |
Gene Name: | TBC1 domain family member 22A |
Gene Aliases: |
C22orf4, HSC79E021 |
Location: |
Chr.22:46762617-47224975 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Set Membership: |
|

Genomic Map