Assay Details
Target Gene Details
Entrez Gene ID: | 105373133 |
Gene Name: | uncharacterized LOC105373133 |
Gene Aliases: |
- |
Location: |
Chr.X:13044544-13086975 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373133 | XM_011545614.2 | XP_011543916.1 |

Genomic Map