Assay Details
Target Gene Details
Entrez Gene ID: | 4128 |
Gene Name: | monoamine oxidase A |
Gene Aliases: |
MAO-A |
Location: |
Chr.X:43654907-43746824 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOA | NM_000240.3 | NP_000231.1 | ||
| NM_001270458.1 | NP_001257387.1 | |||
| AK223499.1 | BAD97219.1 | |||
| AK291769.1 | ||||
| AK293926.1 | ||||
| BC008064.2 | AAH08064.1 | |||
| BC044787.1 | AAH44787.1 | |||
| BT006651.1 | AAP35297.1 | |||
| BX501303.1 | ||||
| M68840.1 | AAA59548.1 | |||
| M69226.1 | AAA59549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517873 | Chr.X:43545579 - 43678961 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map