Assay Details
Target Gene Details
Entrez Gene ID: | 5613 |
Gene Name: | protein kinase, X-linked |
Gene Aliases: |
PKX1 |
Location: |
Chr.X:3604343-3713634 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKX | NM_005044.4 | NP_005035.1 | ||
| BC041073.1 | AAH41073.1 | |||
| BE675757.1 | ||||
| X85545.1 | CAA59733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6780 | Chr.X:3630643 - 3656591 on Build GRCh38 | Insertion |
|
| esv3558553 | Chr.X:3643595 - 3643726 on Build GRCh38 | Deletion |
|
| nsv508749 | Chr.X:3638143 - 3647196 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map