Assay Details
Target Gene Details
Entrez Gene ID: | 9185 |
Gene Name: | RALBP1 associated Eps domain containing 2 |
Gene Aliases: |
POB1 |
Location: |
Chr.X:16946691-17194274 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REPS2 | NM_001080975.1 | NP_001074444.1 | ||
| NM_004726.2 | NP_004717.2 | |||
| XM_005274625.3 | XP_005274682.1 | |||
| XM_005274626.3 | XP_005274683.1 | |||
| XM_011545603.2 | XP_011543905.1 | |||
| XM_011545604.2 | XP_011543906.1 | |||
| XM_011545605.2 | XP_011543907.1 | |||
| XM_011545606.2 | XP_011543908.1 | |||
| XM_011545607.2 | XP_011543909.1 | |||
| XM_017029955.1 | XP_016885444.1 | |||
| XM_017029956.1 | XP_016885445.1 | |||
| XM_017029957.1 | XP_016885446.1 | |||
| XM_017029958.1 | XP_016885447.1 | |||
| AF010233.1 | AAC02901.1 | |||
| AF511533.1 | AAM43933.1 | |||
| AF512951.1 | AAM43953.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs200749769] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map