Assay Details
Target Gene Details
Entrez Gene ID: | 2892 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 3 |
Gene Aliases: |
GLUR-C, GLUR-K3, GLUR3, GLURC, GluA3, MRX94 |
Location: |
Chr.X:123184243-123490915 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIA3 | NM_000828.4 | NP_000819.3 | ||
| NM_001256743.1 | NP_001243672.1 | |||
| NM_007325.4 | NP_015564.4 | |||
| AK297136.1 | ||||
| BC032004.1 | AAH32004.1 | |||
| BC117464.1 | ||||
| DA191279.1 | ||||
| U10301.1 | AAA67922.1 | |||
| U10302.1 | AAA67923.1 |
More Information
Set Membership: |
|

Genomic Map