Assay Details
Target Gene Details
Entrez Gene ID: | 23676 |
Gene Name: | small muscle protein, X-linked |
Gene Aliases: |
DFN6, DFNX4 |
Location: |
Chr.X:21705972-21758160 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPX | NM_014332.2 | NP_055147.1 | ||
| NR_045617.1 | ||||
| AF129505.1 | AAF19343.1 | |||
| AJ250584.1 | CAC08492.1 | |||
| BC005948.1 | AAH05948.1 | |||
| BF126275.1 | ||||
| BM674432.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs151247764] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map