Assay Details
Target Gene Details
Entrez Gene ID: | 55841 |
Gene Name: | WWC family member 3 |
Gene Aliases: |
BM042 |
Location: |
Chr.X:10015755-10144478 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC3 | NM_015691.3 | NP_056506.2 | ||
| AI306162.1 | ||||
| BU500300.1 | ||||
| BU500742.1 | ||||
| KC987947.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113286430] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map