Assay Details
Target Gene Details
Entrez Gene ID: | 159091 |
Gene Name: | family with sequence similarity 122C |
Gene Aliases: |
- |
Location: |
Chr.X:134796413-134855137 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 159091 |
Gene Name: | family with sequence similarity 122C |
Gene Aliases: |
- |
Location: |
Chr.X:134796413-134855137 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |