Assay Details
Target Gene Details
Entrez Gene ID: | 349408 |
Gene Name: | TLR8 antisense RNA 1 |
Gene Aliases: |
GS1-324M7.6 |
Location: |
Chr.X:12902817-12943300 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLR8-AS1 | NR_030727.1 | 5 | 907 |

Genomic Map