Assay Details
Target Gene Details
Entrez Gene ID: | 85417 |
Gene Name: | cyclin B3 |
Gene Aliases: |
- |
Location: |
Chr.X:50202713-50351914 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNB3 | XM_017029913.1 | XP_016885402.1 | ||
| XM_017029914.1 | XP_016885403.1 | |||
| XM_017029915.1 | XP_016885404.1 | |||
| XM_017029916.1 | XP_016885405.1 | |||
| XM_017029917.1 | XP_016885406.1 | |||
| XM_017029918.1 | XP_016885407.1 | |||
| AJ421672.1 | CAD18845.1 | |||
| AJ421673.1 | CAD18846.1 | |||
| AJ421674.1 | CAD18847.1 | |||
| AK292454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv513763 | Chr.X:50206846 - 50209324 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map