Assay Details
Target Gene Details
Entrez Gene ID: | 101928389 |
Gene Name: | uncharacterized LOC101928389 |
Gene Aliases: |
- |
Location: |
Chr.X:17528435-17587160 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928389 | NR_135631.1 |
Target Gene Details
Entrez Gene ID: | 4810 |
Gene Name: | NHS actin remodeling regulator |
Gene Aliases: |
CTRCT40, CXN, SCML1 |
Location: |
Chr.X:17375420-17735994 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHS | NM_001291867.1 | NP_001278796.1 | ||
| NM_198270.3 | NP_938011.1 | |||
| AK302437.1 | ||||
| AY436752.1 | AAR03104.1 | |||
| BC136415.1 | ||||
| BC171763.1 | ||||
| GQ988776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525786 | Chr.X:17453781 - 17542854 on Build GRCh38 | Gain |
|
| dgv503n21 | Chr.X:17329494 - 17542854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map