Assay Details
Target Gene Details
Entrez Gene ID: | 170685 |
Gene Name: | nudix hydrolase 10 |
Gene Aliases: |
APS2, DIPP3-alpha, DIPP3a |
Location: |
Chr.X:51332231-51337525 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT10 | NM_153183.3 | NP_694853.1 | ||
| AF469196.1 | AAM64113.1 |
More Information
Set Membership: |
|

Genomic Map