Assay Details
Target Gene Details
Entrez Gene ID: | 23229 |
Gene Name: | Cdc42 guanine nucleotide exchange factor 9 |
Gene Aliases: |
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2 |
Location: |
Chr.X:63634967-63786025 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
More Information
Set Membership: |
|

Genomic Map