Assay Details
Target Gene Details
Entrez Gene ID: | 105373133 |
Gene Name: | uncharacterized LOC105373133 |
Gene Aliases: |
- |
Location: |
Chr.X:13044544-13086975 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373133 | XM_011545614.2 | XP_011543916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6804 | Chr.X:13059395 - 13088133 on Build GRCh38 | Insertion |
|
| esv3576773 | Chr.X:13071156 - 13600068 on Build GRCh38 | Gain |
|

Genomic Map