Assay Details
Target Gene Details
Entrez Gene ID: | 104798195 |
Gene Name: | uncharacterized LOC104798195 |
Gene Aliases: |
- |
Location: |
Chr.X:15602881-15621484 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GS1-594A7.3 | NR_126564.1 | 2 | 423 | |
| BQ013720.1 | 1 | 167 |

Genomic Map