Assay Details
Target Gene Details
Entrez Gene ID: | 395 |
Gene Name: | Rho GTPase activating protein 6 |
Gene Aliases: |
RHOGAP6, RHOGAPX-1 |
Location: |
Chr.X:11137543-11665701 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP6 | NM_001287242.1 | NP_001274171.1 | ||
| NM_006125.2 | NP_006116.2 | |||
| NM_013423.2 | NP_038267.1 | |||
| NM_013427.2 | NP_038286.2 | |||
| NR_109776.1 | ||||
| AB208792.1 | BAD92029.1 | |||
| AF012272.2 | AAC98539.2 | |||
| AF022212.2 | AAC98540.2 | |||
| AF117067.1 | AAF43261.1 | |||
| AF177663.1 | AAD53166.1 | |||
| AF177665.1 | AAD55087.1 | |||
| AK293078.1 | ||||
| AK297713.1 | ||||
| AK297747.1 | ||||
| AK303444.1 | ||||
| BC150635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515163 | Chr.X:11128177 - 11277520 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map