Assay Details
Target Gene Details
Entrez Gene ID: | 23229 |
Gene Name: | Cdc42 guanine nucleotide exchange factor 9 |
Gene Aliases: |
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2 |
Location: |
Chr.X:63634967-63786025 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF9 | XM_005262249.2 | XP_005262306.1 | ||
| XM_017029369.1 | XP_016884858.1 | |||
| XM_017029378.1 | XP_016884867.1 |
Target Gene Details
Entrez Gene ID: | 100302115 |
Gene Name: | microRNA 1468 |
Gene Aliases: |
MIRN1468, hsa-mir-1468, mir-1468 |
Location: |
Chr.X:63786002-63786087 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR1468 | NR_031567.1 |
More Information
Set Membership: |
|

Genomic Map