Assay Details
Target Gene Details
Entrez Gene ID: | 55841 |
Gene Name: | WWC family member 3 |
Gene Aliases: |
BM042 |
Location: |
Chr.X:10015755-10144478 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC3 | NM_015691.3 | NP_056506.2 | ||
| AI306162.1 | ||||
| BU500300.1 | ||||
| BU500742.1 | ||||
| KC987947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25793 | Chr.X:10014469 - 10017791 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map