Assay Details
Target Gene Details
Entrez Gene ID: | 538 |
Gene Name: | ATPase copper transporting alpha |
Gene Aliases: |
DSMAX, MK, MNK, SMAX3 |
Location: |
Chr.X:77910656-78050395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP7A | NM_000052.6 | NP_000043.4 | ||
| NM_001282224.1 | NP_001269153.1 | |||
| NR_104109.1 | ||||
| AB117973.1 | BAC82353.1 | |||
| AB208828.1 | BAD92065.1 | |||
| AK096667.1 | ||||
| BP364137.1 | ||||
| DB164588.1 | ||||
| L06133.1 | AAA35580.1 |
More Information
Set Membership: |
|

Genomic Map