Assay Details
Target Gene Details
Entrez Gene ID: | 5613 |
Gene Name: | protein kinase, X-linked |
Gene Aliases: |
PKX1 |
Location: |
Chr.X:3604343-3713634 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKX | NM_005044.4 | NP_005035.1 | ||
| BC041073.1 | AAH41073.1 | |||
| X85545.1 | CAA59733.1 |
More Information
Set Membership: |
|

Genomic Map