Assay Details
Target Gene Details
Entrez Gene ID: | 154796 |
Gene Name: | angiomotin |
Gene Aliases: |
- |
Location: |
Chr.X:112774877-112840946 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMOT | NM_001113490.1 | NP_001106962.1 | ||
| NM_133265.2 | NP_573572.1 | |||
| XM_005262087.1 | XP_005262144.1 | |||
| XM_005262090.1 | XP_005262147.1 | |||
| XM_011530875.2 | XP_011529177.1 | |||
| XM_017029289.1 | XP_016884778.1 | |||
| AB028994.3 | BAA83023.3 | |||
| AF286598.1 | AAG01851.1 | |||
| AY987378.1 | AAY24451.1 | |||
| BC130294.1 |
More Information
Set Membership: |
|

Genomic Map