Assay Details
Target Gene Details
Entrez Gene ID: | 2710 |
Gene Name: | glycerol kinase |
Gene Aliases: |
GK1, GKD |
Location: |
Chr.X:30653348-30731462 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 2710 |
Gene Name: | glycerol kinase |
Gene Aliases: |
GK1, GKD |
Location: |
Chr.X:30653348-30731462 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Set Membership: |
|